Anti-BCL7C Mouse Monoclonal Antibody [clone: 5F1]
ABNOH00009274-M01
: Abnova
- Antibody type:Primary
- Antigen name:B-Cell CLL/Lymphoma 7C
- Antigen symbol:BCL7C
- Clonality:Monoclonal
- Clone:5F1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9274
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: BCL7C
Clonality: Monoclonal
Clone: 5F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human