Anti-ITGB1BP1 Mouse Polyclonal Antibody
Catalog # ABNOH00009270-A01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Integrin Beta 1 Binding Protein 1
- Antigen symbol:ITGB1BP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9270
- Antigen synonyms:ICAP-1B|ICAP1|ITGB1BP|CHORDC3|ICAP-1alpha|ICAP1A|ICAP1B|MSTP015|ICAP-1A|MELUSIN
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: ITGB1BP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human