Anti-NRP1 Mouse Monoclonal Antibody [clone: 1B3]
ABNOH00008829-M05
: Abnova
- Antibody type:Primary
- Antigen name:Neuropilin 1
- Antigen symbol:NRP1
- Clonality:Monoclonal
- Clone:1B3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:8829
- Antigen synonyms:NP1|VEGF165R|BDCA4|NRP|CD304
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: NRP1
Clonality: Monoclonal
Clone: 1B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human