Anti-PEX11B Mouse Monoclonal Antibody [clone: 2D2]
Catalog # ABNOH00008799-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Peroxisomal Biogenesis Factor 11 beta
- Antigen symbol:PEX11B
- Clonality:Monoclonal
- Clone:2D2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:8799
- Antigen synonyms:Peroxin 11 beta|Protein PEX11 homolog beta|Peroxisomal biogenesis factor 11B|Peroxin-11 beta|PEX11B|PEX11-beta|Peroxin 11B|Peroxisomal membrane protein 11B|PEX-11 beta|Peroxin11B|PEX11 beta
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PEX11B
Clonality: Monoclonal
Clone: 2D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human