Anti-NOP14 Mouse Polyclonal Antibody
Catalog # ABNOH00008602-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:NOP14 nucleolar Protein
- Antigen symbol:NOP14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:8602
- Antigen synonyms:RES4-25|RES425|UTP2|C4orf9|NOL14
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: NOP14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human