Anti-EEA1 Mouse Polyclonal Antibody
Catalog # ABNOH00008411-A01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Early Endosome Antigen 1
- Antigen symbol:EEA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:8411
- Antigen synonyms:ZFYVE2|MSTP105|MST105
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: EEA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat