Anti-ZNF79 Mouse Monoclonal Antibody [clone: 3D12]
Catalog # ABNOH00007633-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Zinc Finger Protein 79
- Antigen symbol:ZNF79
- Clonality:Monoclonal
- Clone:3D12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:7633
- Antigen synonyms:pT7
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: ZNF79
Clonality: Monoclonal
Clone: 3D12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human