Order Entry
Italy
ContactUsLinkComponent
Anti-ZNF76 Mouse Monoclonal Antibody [clone: 3D4]
Anti-ZNF76 Mouse Monoclonal Antibody [clone: 3D4]
Catalog # ABNOH00007629-M06
Supplier:  Abnova
undefined
Anti-ZNF76 Mouse Monoclonal Antibody [clone: 3D4]
Catalog # ABNOH00007629-M06
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Zinc Finger Protein 76
  • Antigen symbol:
    ZNF76
  • Clonality:
    Monoclonal
  • Clone:
    3D4
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    7629
  • Antigen synonyms:
    Zfp523|D6S229E|ZNF523
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: ZNF76
Clonality: Monoclonal
Clone: 3D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human