Anti-ZNF69 Mouse Monoclonal Antibody [clone: 1E3]
Catalog # ABNOH00007620-M08
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Zinc Finger Protein 69
- Antigen symbol:ZNF69
- Clonality:Monoclonal
- Clone:1E3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:7620
- Antigen synonyms:Cos5
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: ZNF69
Clonality: Monoclonal
Clone: 1E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human