Anti-UCHL1 Mouse Monoclonal Antibody [clone: S1]
Catalog # ABNOH00007345-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ubiquitin Carboxyl-Terminal Esterase L1
- Antigen symbol:UCHL1
- Clonality:Monoclonal
- Clone:S1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:7345
- Antigen synonyms:Neuron cytoplasmic protein 9.5|Ubiquitin thioesterase L1.|PGP9.5|Ubiquitin carboxyl-terminal|hydrolase isozyme L1|Ubiquitin thioesterase L1|UCH-L1|PGP 9.5|PARK5
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: UCHL1
Clonality: Monoclonal
Clone: S1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human