Anti-VPS72 Mouse Monoclonal Antibody [clone: 2G6]
Catalog # ABNOH00006944-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Vacuolar Protein Sorting 72
- Antigen symbol:VPS72
- Clonality:Monoclonal
- Clone:2G6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:6944
- Antigen synonyms:Swc2|YL1|TCFL1|YL-1|CFL1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: VPS72
Clonality: Monoclonal
Clone: 2G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human