Anti-SNAPC4 Mouse Monoclonal Antibody [clone: 3F5]
ABNOH00006621-M08
: Abnova
- Antibody type:Primary
- Antigen name:Small Nuclear RNA Activating Complex, Polypeptide 4, 190kDa
- Antigen symbol:SNAPC4
- Clonality:Monoclonal
- Clone:3F5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:6621
- Antigen synonyms:SNAP190|PTFalpha
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: SNAPC4
Clonality: Monoclonal
Clone: 3F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human