Anti-GRK1 Mouse Monoclonal Antibody [clone: 4E9]
ABNOH00006011-M02A
: Abnova
- Antibody type:Primary
- Antigen name:G Protein-Coupled Receptor Kinase 1
- Antigen symbol:GRK1
- Clonality:Monoclonal
- Clone:4E9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:6011
- Antigen synonyms:RK|Rhodopsin kinase|GPRK1|RHOK
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: GRK1
Clonality: Monoclonal
Clone: 4E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human