Anti-PSMC3 Rabbit Polyclonal Antibody
Catalog # ABNOH00005702-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Proteasome 26S Subunit, ATPase 3
- Antigen symbol:PSMC3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5702
- Antigen synonyms:TBP1|macropain) 26S Subunit|proteasome 26S subunit|Proteasome (prosome|ATPase 3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PSMC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human