Anti-PRKAB2 Rabbit Polyclonal Antibody
Catalog # ABNOH00005565-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Protein Kinase, AMP-activated Non-catalytic Subunit Beta 2
- Antigen symbol:PRKAB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5565
- Antigen synonyms:BB124140|5730553K21Rik|AW049591
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PRKAB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse