Anti-PRKAB2 Mouse Polyclonal Antibody
Catalog # ABNOH00005565-A01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Protein Kinase, AMP-activated Non-catalytic Subunit Beta 2
- Antigen symbol:PRKAB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5565
- Antigen synonyms:BB124140|5730553K21Rik|AW049591
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: PRKAB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human