Anti-PRKAA2 Mouse Monoclonal Antibody [clone: 1G8]
Catalog # ABNOH00005563-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Protein Kinase AMP-activated Catalytic Subunit Alpha 2
- Antigen symbol:PRKAA2
- Clonality:Monoclonal
- Clone:1G8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- ChIP:Yes
- Gene ID:5563
- Antigen synonyms:AMPKa2|AMPK|AMPK2|PRKAA
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PRKAA2
Clonality: Monoclonal
Clone: 1G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human