Anti-PPP6C Mouse Monoclonal Antibody [clone: 1B7]
ABNOH00005537-M01
: Abnova
- Antibody type:Primary
- Antigen name:Protein Phosphatase 6, Catalytic Subunit
- Antigen symbol:PPP6C
- Clonality:Monoclonal
- Clone:1B7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:5537
- Antigen synonyms:PP6C|Serine/threonine protein phosphatase 6|PP6
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: PPP6C
Clonality: Monoclonal
Clone: 1B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human