Anti-PFKFB3 Mouse Polyclonal Antibody
Catalog # ABNOH00005209-A01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 3
- Antigen symbol:PFKFB3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5209
- Antigen synonyms:6PF-2-K/Fru-2,6-P2ASE brain/placenta-type isozyme|Fructose-2,6-bisphosphatase|iPFK-2|6-phosphofructo-2-kinase|IPFK2|PFK2|6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: PFKFB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human