Anti-PCDH8 Mouse Monoclonal Antibody [clone: 6A8]
Catalog # ABNOH00005100-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Protocadherin 8
- Antigen symbol:PCDH8
- Clonality:Monoclonal
- Clone:6A8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- ChIP:Yes
- Gene ID:5100
- Antigen synonyms:Arcadlin|PAPC|Protocadherin8|Protocadherin 8|PCDH 8|PCDH8_HUMAN.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PCDH8
Clonality: Monoclonal
Clone: 6A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human