Anti-PAFAH2 Mouse Monoclonal Antibody [clone: 1A8]
Catalog # ABNOH00005051-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Platelet-activating Factor Acetylhydrolase 2
- Antigen symbol:PAFAH2
- Clonality:Monoclonal
- Clone:1A8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:5051
- Antigen synonyms:HSD-PLA2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PAFAH2
Clonality: Monoclonal
Clone: 1A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human