Order Entry
Italy
ContactUsLinkComponent
Anti-NDUFA8 Mouse Monoclonal Antibody [clone: 2E10]
Anti-NDUFA8 Mouse Monoclonal Antibody [clone: 2E10]
Catalog # ABNOH00004702-M05
Supplier:  Abnova
undefined
Anti-NDUFA8 Mouse Monoclonal Antibody [clone: 2E10]
Catalog # ABNOH00004702-M05
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    NADH dehydrogenase (ubiquinone) 1, alpha subcomplex 8
  • Antigen symbol:
    NDUFA8
  • Clonality:
    Monoclonal
  • Clone:
    2E10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    4702
  • Antigen synonyms:
    NDUFA8|CI-PGIV|NADH-ubiquinone oxidoreductase 19 kDa subunit|CI-19kD|Complex I-PGIV|Complex I-19kD|NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 8
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: NDUFA8
Clonality: Monoclonal
Clone: 2E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human