Anti-SH2D1A Mouse Monoclonal Antibody [clone: 1C9]
ABNOH00004068-M01
: Abnova
- Antibody type:Primary
- Antigen name:SH2 domain containing 1A
- Antigen symbol:SH2D1A
- Clonality:Monoclonal
- Clone:1C9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:4068
- Antigen synonyms:RP5-1052M9.3|FLJ18687|IMD5|EBVS|signaling lymphocyte activation molecule-associated protein|FLJ92177|SH2D1A|XLP|SLAM (signaling lymphocyte activation molecule) associated protein|SLAM-associated protein|HGNC:10820|SAP|XLPD |SH2 domain protein 1A|MTCP1|T cell signal transduction molecule SAP|LYP|Duncan's disease (lymphoproliferative syndrome)|DSHP
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: SH2D1A
Clonality: Monoclonal
Clone: 1C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human