Anti-HOXB9 Mouse Monoclonal Antibody [clone: 2E9]
Catalog # ABNOH00003219-M06
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Homeobox B9
- Antigen symbol:HOXB9
- Clonality:Monoclonal
- Clone:2E9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:3219
- Antigen synonyms:HOX-2.5|HOX2|HOX2E
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: HOXB9
Clonality: Monoclonal
Clone: 2E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human