Anti-HOXA5 Mouse Monoclonal Antibody [clone: 4A8]
Catalog # ABNOH00003202-M05
Nome del fornitore: Abnova
Specifiche
- Antibody type:Primary
- Antigen name:Homeobox A5
- Antigen symbol:HOXA5
- Clonality:Monoclonal
- Clone:4A8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:3202
- Antigen synonyms:Homeo box 1C Homeobox A5 Homeobox protein HOXA5 HOX 1 HOX 1.3 HOX 1C HOX1 HOX1.3 HOX1C|HXA5_HUMAN.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifiche
Informazioni su questo articolo
Type: Primary
Antigen: HOXA5
Clonality: Monoclonal
Clone: 4A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human