Anti-HLA-DMB Mouse Monoclonal Antibody [clone: 4G8]
Catalog # ABNOH00003109-M04
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:HLA class II histocompatibility antigen, DM beta chain
- Antigen symbol:HLA-DMB
- Clonality:Monoclonal
- Clone:4G8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:3109
- Antigen synonyms:D6S221E|RING7
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: HLA-DMB
Clonality: Monoclonal
Clone: 4G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human