Anti-EN2 Mouse Monoclonal Antibody [clone: 1E1]
ABNOH00002020-M03
: Abnova
- Antibody type:Primary
- Antigen name:Engrailed 2
- Antigen symbol:EN2
- Clonality:Monoclonal
- Clone:1E1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2020
- Antigen synonyms:Hu-En-2 |EN2|Homeobox protein engrailed-2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: EN2
Clonality: Monoclonal
Clone: 1E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human