Anti-TIMM8A Mouse Monoclonal Antibody [clone: 2D12]
Catalog # ABNOH00001678-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Translocase of Inner Mitochondrial Membrane 8 homolog A
- Antigen symbol:TIMM8A
- Clonality:Monoclonal
- Clone:2D12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:1678
- Antigen synonyms:TIM8|DFN1|DDP|MTS|DDP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: TIMM8A
Clonality: Monoclonal
Clone: 2D12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human