Anti-AP2S1 Mouse Monoclonal Antibody [clone: 3E4]
Catalog # ABNOH00001175-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Adaptor-Related Protein Complex 2, Sigma 1 Subunit
- Antigen symbol:AP2S1
- Clonality:Monoclonal
- Clone:3E4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- ChIP:Yes
- Gene ID:1175
- Antigen synonyms:HHC3|FBH3|AP17|CLAPS2|FBHOk
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: AP2S1
Clonality: Monoclonal
Clone: 3E4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human