Anti-CEL Mouse Monoclonal Antibody [clone: 3C8]
ABNOH00001056-M15
: Abnova
- Antibody type:Primary
- Antigen name:Carboxyl ester lipase
- Antigen symbol:CEL
- Clonality:Monoclonal
- Clone:3C8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:1056
- Antigen synonyms:FAP|BSDL|FAPP|MODY8|LIPA|BAL|BSSL|CEase|CELL
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: CEL
Clonality: Monoclonal
Clone: 3C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human