Anti-CD69 Mouse Monoclonal Antibody [clone: 4H3]
Catalog # ABNOH00000969-M07
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:CD69 molecule
- Antigen symbol:CD69
- Clonality:Monoclonal
- Clone:4H3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Cross adsorption:No
- Gene ID:969
- Antigen synonyms:EA1|MLR-3|AIM|GP32/28|CLEC2C|BL-AC/P26
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: 4H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human