Order Entry
Italy
ContactUsLinkComponent
Anti-CA8 Mouse Monoclonal Antibody [clone: 1F7]
Anti-CA8 Mouse Monoclonal Antibody [clone: 1F7]
Catalog # ABNOH00000767-M01
Supplier:  Abnova
undefined
Anti-CA8 Mouse Monoclonal Antibody [clone: 1F7]
Catalog # ABNOH00000767-M01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Carbonic Anhydrase 8
  • Antigen symbol:
    CA8
  • Clonality:
    Monoclonal
  • Clone:
    1F7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    767
  • Antigen synonyms:
    CALS|CA-VIII|CARP|CAMRQ3
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: CA8
Clonality: Monoclonal
Clone: 1F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human