Anti-CA3 Mouse Monoclonal Antibody [clone: 4A12-1A3]
Catalog # ABNOH00000761-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Carbonic Anhydrase 3
- Antigen symbol:CA3
- Clonality:Monoclonal
- Clone:4A12-1A3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:761
- Antigen synonyms:CAIII|Car3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CA3
Clonality: Monoclonal
Clone: 4A12-1A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human