Anti-CA1 Mouse Monoclonal Antibody [clone: 1F1]
Catalog # ABNOH00000759-M10
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Carbonic Anhydrase 1
- Antigen symbol:CA1
- Clonality:Monoclonal
- Clone:1F1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:759
- Antigen synonyms:Carbonic anhydrase 1|Carbonic dehydratase|HEL-S-11|Car1|CA-I|CAB|Carbonate dehydratase I|ECK0125|yadF|Carbonic anhydrase B
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CA1
Clonality: Monoclonal
Clone: 1F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human