Anti-POLR3D Mouse Monoclonal Antibody [clone: 4E6]
Catalog # ABNOH00000661-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Polymerase (RNA) III (DNA directed) polypeptide D (44kDa)
- Antigen symbol:POLR3D
- Clonality:Monoclonal
- Clone:4E6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:661
- Antigen synonyms:RPC4|BN51T|TSBN51|RPC53
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: POLR3D
Clonality: Monoclonal
Clone: 4E6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human