Specifications
- Antibody type:Primary
- Antigen name:BMI1 Proto-Oncogene, Polycomb Ring Finger
- Antigen symbol:BMI1
- Clonality:Monoclonal
- Clone:4E10-1C5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:648
- Antigen synonyms:ring finger protein 51|murine leukemia viral (bmi-1) oncogene homolog|flvi-2/bmi-1|polycomb group RING finger protein 4|polycomb group protein Bmi1|RNF51|B lymphoma Mo-MLV insertion region 1 homolog|FLVI2/BMI1|PCGF4|RP11-573G6.1|polycomb complex protein BMI-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: BMI1
Clonality: Monoclonal
Clone: 4E10-1C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human