Order Entry
Italy
ContactUsLinkComponent
 
Anti-ADGRB2 Mouse Monoclonal Antibody [clone: 6A12]
  ABNOH00000576-M01
 :  Abnova
undefined
Anti-ADGRB2 Mouse Monoclonal Antibody [clone: 6A12]
  ABNOH00000576-M01
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Adhesion G Protein-Coupled Receptor B2
  • Antigen symbol:
    ADGRB2
  • Clonality:
    Monoclonal
  • Clone:
    6A12
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    576
  • Antigen synonyms:
    BAI2|Brain-specific angiogenesis inhibitor 2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: ADGRB2
Clonality: Monoclonal
Clone: 6A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human