Anti-ATP6V1B1 Mouse Monoclonal Antibody [clone: 3G11]
ABNOH00000525-M02
: Abnova
- Antibody type:Primary
- Antigen name:ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B1
- Antigen symbol:ATP6V1B1
- Clonality:Monoclonal
- Clone:3G11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:525
- Antigen synonyms:RTA1B|ATP6B1|VATB|VMA2|VPP3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: ATP6V1B1
Clonality: Monoclonal
Clone: 3G11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human