Anti-RHOC Mouse Monoclonal Antibody [clone: 2E12]
Catalog # ABNOH00000389-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ras Homolog Family Member C
- Antigen symbol:RHOC
- Clonality:Monoclonal
- Clone:2E12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:389
- Antigen synonyms:MGC1448|MGC61427|H9|ARH9|ARHC|RHOH9
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: RHOC
Clonality: Monoclonal
Clone: 2E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat