Anti-MKI67 Mouse Monoclonal Antibody [clone: OTI1F11]
ORIGTA802426
: OriGene
- Antibody type:Primary
- Antigen name:Marker of proliferation Ki-67
- Antigen symbol:MKI67
- Clonality:Monoclonal
- Clone:OTI1F11
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Amino acid number:1160 to 1493
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Protein
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Immunochemistry: 1:100; Western Blot: 1:200-1:1000
Type: Primary
Antigen: MKI67 (marker of proliferation Ki-67)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human