Anti-PYCR2 Mouse Monoclonal Antibody [clone: OTI2F11]
Catalog # ORIGTA501898
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Pyrroline-5-carboxylate reductase 2
- Antigen symbol:PYCR2
- Clonality:Monoclonal
- Clone:OTI2F11
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:33.5 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100
Type: Primary
Antigen: PYCR2 (pyrroline-5-carboxylate reductase family, member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human