Anti-SLC7A8 Mouse Monoclonal Antibody [clone: OTI7D6]
Catalog # ORIGTA500515
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 7 (amino acid transporter light chain, L system), member 8
- Antigen symbol:Solute carrier family 7, member 8
- Clonality:Monoclonal
- Clone:OTI7D6
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:58.4 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50
Type: Primary
Antigen: SLC7A8 (solute carrier family 7 (amino acid transporter light chain, L system), member 8)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human