Anti-CDK11A Rabbit Polyclonal Antibody
Catalog # ORIGTA316484
Supplier: OriGene
Jump to:
Product Details & Documents
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000
Type: Primary
Antigen: CDK11A (cyclin-dependent kinase 11A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Documents
TA316484.pdf
Datasheet
Specifications
- Western blot:Yes
- Clonality:Polyclonal
- Purification:purified by affinity chromatography
- Antibody type:Primary
- Epitope:internal
- Form:Liquid
- Isotype:IgG
- Storage temperature:Cool
- Host:Rabbit
- Cross adsorption:none
- Shipping temperature:Ice
- Antigen name:Cyclin-dependent kinase 11A
- Cat. No.:ORIGTA316484
- Reactivity:Human
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:CDK11A
- Storage buffer:Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.



