Anti-PRKAA2 Goat Antibody
Catalog # ORIGTA311358
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Protein kinase, AMP-activated, alpha 2 catalytic subunit
- Antigen symbol:PRKAA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Form:Liquid
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.Aliquot and store at -20C. Minimize freezing and thawing.
- Sequence:CPLDALNTTKP
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by ammonium sulphate precipitation followed by antigen affinity chromatography
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1-3?g/ml; ELISA: 1.5-3?g/ml
Type: Primary
Antigen: PRKAA2 (protein kinase, AMP-activated, alpha 2 catalytic subunit)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human