Order Entry
Italy
ContactUsLinkComponent
Anti-CASP3 Mouse monoclonal antibody [clone: CPP32 4-1-18]
Anti-CASP3 Mouse monoclonal antibody [clone: CPP32 4-1-18]
Catalog # ORIGTA301776
Supplier:  OriGene
Anti-CASP3 Mouse monoclonal antibody [clone: CPP32 4-1-18]
Catalog # ORIGTA301776
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    caspase 3, apoptosis-related cysteine peptidase
  • Antigen symbol:
    CASP3
  • Clonality:
    Monoclonal
  • Clone:
    CPP32 4-1-18
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    Full length
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    0,1% sodium azide
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography
  • Pk:
    200 µl

Specifications

About this item

Recommended Dilutions: Western Blot: 1:500-1: 1000

Type: Primary
Antigen: CASP3 (caspase 3, apoptosis-related cysteine peptidase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human