Anti-SLC5A7 Mouse Monoclonal Antibody [clone: 62-2E8]
Catalog # ORIGTA301538
Nome del fornitore: OriGene
Specifiche
- Antibody type:Primary
- Antigen name:solute carrier family 5 (sodium/choline cotransporter), member 7
- Antigen symbol:Solute carrier family 5, member 7
- Clonality:Monoclonal
- Clone:62-2E8
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Format:Ascites
- Form:Liquid
- Storage buffer:PBS and 0,05% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Protein
- Pk:100 µl
Specifiche
Informazioni su questo articolo
Recommended Dilutions: Western Blot: 1: 1000
Type: Primary
Antigen: SLC5A7 (solute carrier family 5 (sodium/choline cotransporter), member 7)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat