Anti-ZsGreen1 Mouse Monoclonal Antibody
ORIGTA180002
: OriGene
- Antibody type:Tag
- Antigen symbol:ZsGreen1
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Type: Tag
Antigen: ZsGreen1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: