Order Entry
Italy
ContactUsLinkComponent
Anti-PTPRC Rat Monoclonal Antibody [clone: 30-F11]
Anti-PTPRC Rat Monoclonal Antibody [clone: 30-F11]
Catalog # 70-0451-U100
Supplier:  Tonbo Biosciences
undefined
Anti-PTPRC Rat Monoclonal Antibody [clone: 30-F11]
Catalog # 70-0451-U100
Supplier:  Tonbo Biosciences

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein tyrosine phosphatase receptor type C
  • Antigen symbol:
    PTPRC
  • Clonality:
    Monoclonal
  • Clone:
    30-F11
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Gene ID:
    19264
  • Antigen synonyms:
    CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µG

Specifications

About this item

The 30-F11 antibody reacts with mouse CD45, which is one of the most abundant hematopoietic markers and is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). CD45 is a protein tyrosine phosphatase existing in several isoforms, each being generated and expressed in cell-specific patterns. With its broad cell distribution, CD45 is critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, include CD45R (B220), CD45RA, CD45RB and CD45RO.

The 30-F11 antibody is widely used as a leukocyte marker for B cells, T cell subsets and NK cell subsets.

Type: Primary
Antigen: PTPRC (protein tyrosine phosphatase, receptor type, C)
Clonality: monoclonal
Clone: 30-F11
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity: Mouse