Anti-HLA-DRB5 Rabbit Polyclonal Antibody
ABGEAP9825C
: Abgent
- Antibody type:Primary
- Antigen name:HLA class II histocompatibility antigen, DRB5 beta chain
- Antigen symbol:HLA-DRB5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:Center
- Gene ID:284366
- Antigen synonyms:HLA-DRB
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:PGFNKDLSANDHNDDC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This KLK9 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 82-111 amino acids from the Central region of human KLK9.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: HLA-DRB5 (major histocompatibility complex, class II, DR beta 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human