Anti-MAT2B Rabbit Polyclonal Antibody
ABGEAP9534A
: Abgent
- Antibody type:Primary
- Antigen name:Methionine Adenosyltransferase 2 Beta
- Antigen symbol:MAT2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal
- Gene ID:2034
- Antigen synonyms:SDR23E1|MATIIbeta|MAT-II|TGR|Nbla02999
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:LESKKTEPEHRPMSC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,4 mg/ml
- Shipping temperature:4 °C
- Immunogen:This EPAS1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 577-605 amino acids from the C-terminal region of human EPAS1.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: MAT2B (methionine adenosyltransferase II, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human